Orders LinkContactUsLinkComponent
Calibration weight sets, class E2, Proof Line
Calibration weight sets, class E2, Proof Line
1 mg - 500 mg, Without certificate
Catalog # SARBYCS011-252AC00
Supplier:   Sartorius Balances
Calibration weight sets, class E2, Proof Line
Catalog # SARBYCS011-252AC00
Supplier:  Sartorius Balances
Calibration weight sets, class E2, Proof Line
Calibration weight sets, class E2, Proof Line
1 mg - 500 mg, Without certificate
Price
531,00 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
These class E2 weight sets include aluminium (1 to 5 mg) or nickel silver (10 to 500 mg) or stainless steel (1 g and higher) weights.
Spécifications
  • Weight set:
    1 mg - 500 mg
  • Seq:
    01
  • Type:
    Without certificate