Automatic tilt measures
Sulphuric acid, 10 ml, Clear
N° de catalogue 614-0147
Fournisseur: GERBER FUNKE
Jump to:
Product Details & Documents
Glass, with rubber stopper.
- Superior quality models
- Storage bottle of 500/250 ml
- For milk analysis applications
Colour: Clear
Specifications
- Capacity:10 ml
- Colour:Clear
- Description:Sulphuric acid
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



