ContactUsLinkComponent
Compact, bench-top shaker-incubator. built-in microprocessor thermocontroller provides constant temperature control in the incubato r chamber. forced heated air circulation inside the transparent plexiglass chamber guarantees even temperature distri 1 * 1 items
Catalog # 444-0779
Supplier:  GRANT INSTRUMENTS
Compact, bench-top shaker-incubator. built-in microprocessor thermocontroller provides constant temperature control in the incubato r chamber. forced heated air circulation inside the transparent plexiglass chamber guarantees even temperature distri 1 * 1 items
Catalog # 444-0779
Supplier:  GRANT INSTRUMENTS