Decontamination solution, RNase-ExitusPlus™
RNase-ExitusPlus™, refill pack, 2,5 L
Supplier: PanReac AppliChem
Decontamination solution, RNase-ExitusPlus™
Catalog # A7153.2500RF
Supplier: PanReac AppliChem
CAS Number:
Jump to:
Product Details & Documents
RNase-ExitusPlus™ is a non-alkaline, non-corrosive and non-carcinogenic cleansing solution that is highly active against RNase contamination. RNase-ExitusPlus™ has been demonstrated to inactivate more than 20 μg of RNase A dried onto the bottom of a microcentrifuge tube. RNase-ExitusPlus™ is stable for approximately 18 months and heat resistant. It should be stored at room temperature; at colder temperatures a precipitate may form which is easily brought into solution at 37 °C. RNase-ExitusPlus™ is supplied in spray bottles and a one litre refill pack is also available.
- Catalytic and cooperative effects of the components cause a very rapid non-enzymatic, non-sequence-specific degradation of protein and RNase molecules
- All components of RNase-ExitusPlus™ are readily and not harmful or toxic for humans
- No aggressive mineral acids or alkaline substances are used; equipment and materials are not damaged or corroded even after prolonged incubation times
- No organic solvents or volatile components, no toxic fumes
- Elevated temperatures above approx. 50 °C speed up the reaction and the efficiency
Certificates
Search by Lot NumberApplichem Lifescience Effective DNA and RNA Decontamination in One DNA RNA-ExitusPlusTM Applichem Lifescience ExitusPlus™ – Effective Solution – Effective Decontamination Applichem Lifescience Products for the Investigation of COVID-19 Applichem Lifescience Products Supporting your Work Against COVID-19
Specifications
Specifications
- Pk:2,5 L
- Description:RNase-ExitusPlus™, refill pack



