Chairs
ESD chair, with step, contact backrest with tilting seat, with volume conductive plastic seat and five-legged base frame, 580 - 850 mm, Glides
BIMO9811E-1100
: bimos
These work chairs are user-friendly and ensures flexible sitting at workstations in production. The work chair has a large selection of mechanisms and is equipped as standard with seat and back shells made of sturdy plastic.
- The comfortable change pads are available in different surfaces and colours
- Features exchangeable upholstery elements
- Permanent contact backrest, backrest height adjustment and steel base
- Description:ESD chair, with step, contact backrest with tilting seat, with volume conductive plastic seat and five-legged base frame
- Height:580 - 850 mm
- Seq:17
- Foot style:Glides
Accessories
- Description
- Description
- Description
- Description
- Description
- Description
- Description
- Description
- Description
- Description
- Description