Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Connexion
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
MASK FFP2 WITHOUT VALVE 1 * 200 items
Catalog # VALMVR202V
Supplier:
DELTALYOVALMY
Expand image
MASK FFP2 WITHOUT VALVE 1 * 200 items
Catalog # VALMVR202V
Supplier:
DELTALYOVALMY
Frequently Bought Together
VWR® Nitrile Extra Light, Nitrile Gloves
Supplier: VWR Collection
Nitrile gloves, TouchNTuff® 92-600 / 92-605
Supplier: Ansell
Nitrile gloves, Kimtech™ Purple Nitrile™ Xtra™
Supplier: KIMBERLY CLARK
Gloves, nitrile/neoprene, Microflex® 93-260
Supplier: Ansell
Nitrile gloves, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
VWR®, Nitrile Gloves
Supplier: VWR Collection
pH buffer solutions, NIST traceable, Orion™
Supplier: Thermo Orion
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
VWR®, Laboratory Markers
Supplier: VWR Collection
VWR®, Latex Gloves
Supplier: VWR Collection