Orders LinkContactUsLinkComponent
Objectives
Objectives
EC Epi-Planachromatic, M27, N.A. of 0,8, WD of 1,0 mm
Catalog # ZEIS4220909901000
Supplier:   ZEISS
Objectives
Catalog # ZEIS4220909901000
Supplier:  ZEISS
Objectives
Objectives
EC Epi-Planachromatic, M27, N.A. of 0,8, WD of 1,0 mm
Price
1 478,64 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Magnification:
    100×
  • Description:
    EC Epi-Planachromatic, M27, N.A. of 0,8, WD of 1,0 mm
  • Seq:
    28