Order Entry
Belgium
Orders LinkContactUsLinkComponent
Conductivity cells, IDS

Conductivity cells, IDS

IDS conductivity cell, LR 925/01, 2-cell stainless steel, incl. flow container, LR 925/01, 2-cell stainless steel, incl. flow container, 0,01 - 200 µS/cm, 0,100 cm-1, 0...+100 °C, Stainless steel, 12×120 mm, IDS plug, 1,5 m cable
Catalog # 663-0156
Supplier:   WTW
Conductivity cells, IDS
Catalog # 663-0156
Supplier:  WTW
Conductivity cells, IDS
Conductivity cells, IDS

IDS conductivity cell, LR 925/01, 2-cell stainless steel, incl. flow container, LR 925/01, 2-cell stainless steel, incl. flow container, 0,01 - 200 µS/cm, 0,100 cm-1, 0...+100 °C, Stainless steel, 12×120 mm, IDS plug, 1,5 m cable

Price
999,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

High quality, enhanced sensor technology combined with most up to date measurement electronics. For use with IDS meters.

  • Intelligent: Sensors are automatically recognised by the meter, each sensor is individually identified
  • Digital: Measurement signals are processed digitally, providing very accurate data
  • IDS cells store their serial number and calibration records
  • TetraCon® 925: 4-electrode measuring cell ideal for medium and high conductivity applications
  • LR 925/01: Ideal for pure water and low conductivity applications
  • TetraCon® 925/LV: Graphite 4-electrode cell for small volumes and viscous samples

The electrodes can provide the following data, shown on the screen of the digital meter:
- Model name
- Serial number
- Calibration status (+++, ++, +, -, blank)
- The calibration history includes the last 10 calibrations, including buffers used, mV values, etc.

Ordering information: Various cable lengths available.

Specifications
  • Conductivity:
    0,01 - 200 µS/cm
  • Connection:
    IDS plug, 1,5 m cable
  • Ø×L:
    12×120 mm
  • Type:
    LR 925/01, 2-cell stainless steel, incl. flow container
  • Description:
    IDS conductivity cell, LR 925/01, 2-cell stainless steel, incl. flow container
  • Material:
    Stainless steel
  • Seq:
    06
  • Temp.:
    0...+100 °C
  • Cell constant:
    0,100 cm-1
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.