Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Conductivity cells, IDS

IDS conductivity cell, TetraCon® 925, 4-cell graphite, TetraCon® 925, 4-cell graphite, 10 µS/cm - 2000 mS/cm, 0,475 cm-1, 0...+100 °C, Epoxy, 15,3×120 mm, IDS-plug, 1,5 m cable
Catalog # 663-0152
Supplier:   WTW
Conductivity cells, IDS
Catalog # 663-0152
Supplier:  WTW
Conductivity cells, IDS
Conductivity cells, IDS

IDS conductivity cell, TetraCon® 925, 4-cell graphite, TetraCon® 925, 4-cell graphite, 10 µS/cm - 2000 mS/cm, 0,475 cm-1, 0...+100 °C, Epoxy, 15,3×120 mm, IDS-plug, 1,5 m cable

Price
723,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

High quality, enhanced sensor technology combined with most up to date measurement electronics. For use with IDS meters.

  • Intelligent: Sensors are automatically recognised by the meter, each sensor is individually identified
  • Digital: Measurement signals are processed digitally, providing very accurate data
  • IDS cells store their serial number and calibration records
  • TetraCon® 925: 4-electrode measuring cell ideal for medium and high conductivity applications
  • LR 925/01: Ideal for pure water and low conductivity applications
  • TetraCon® 925/LV: Graphite 4-electrode cell for small volumes and viscous samples

The electrodes can provide the following data, shown on the screen of the digital meter:
- Model name
- Serial number
- Calibration status (+++, ++, +, -, blank)
- The calibration history includes the last 10 calibrations, including buffers used, mV values, etc.

 : Various cable lengths available.

Specifications
  • Conductivity:
    10 µS/cm - 2000 mS/cm
  • Connection:
    IDS-plug, 1,5 m cable
  • Ø×L:
    15,3×120 mm
  • Type:
    TetraCon® 925, 4-cell graphite
  • Description:
    IDS conductivity cell, TetraCon® 925, 4-cell graphite
  • Material:
    Epoxy
  • Seq:
    01
  • Temp.:
    0...+100 °C
  • Cell constant:
    0,475 cm-1