Order Entry
Belgium
Orders LinkContactUsLinkComponent
Capillary tubes

Capillary tubes

Capillary Tubes, 75 mm, 1,75 mm, 82,5 µl
Catalog # MARI2951303
Supplier:   MARIENFELD
Capillary tubes
Catalog # MARI2951303
Supplier:  MARIENFELD
Capillary tubes
Capillary tubes

Capillary Tubes, 75 mm, 1,75 mm, 82,5 µl

Price
Inquire For Price
Log in for availability
1 * 1.000 ST
Product Details & Documents

Glass, clear.

  • Na-heparinized (coated)

Capillaries, Na-heparinized (coated)

For blood gas analysis.

Specifications
  • Ø:
    1,75 mm
  • Volume:
    82,5 µl
  • Description:
    Capillary Tubes
  • Length:
    75 mm
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Closure cap for blood gas capillary tubes with Ø 1,75 mm
    Closure cap for blood gas capillary tubes with Ø 1,75 mm
    MARI2960414
    Log in for availability
    1 * 500 ST

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.