Order Entry
Belgium
Orders LinkContactUsLinkComponent
Labels, pressure-sensitive

Labels, pressure-sensitive

Labels, Polyolefin, White, 20×38 mm
Catalog # 817-0082
   Low Manufacturing Impact
Labels, pressure-sensitive
Catalog # 817-0082
   Low Manufacturing Impact
Labels, pressure-sensitive
Labels, pressure-sensitive

Labels, Polyolefin, White, 20×38 mm

Price
101,00 €
Log in for availability
1 * 650 ST
Product Details & Documents

These Nalgene™ polyolefin pressure-sensitive labels are used with photocopiers, laser printers or inkjet printers. Accepts a variety of inks including copier, marker, ballpoint, gels, and technical markers (pretest specific inks for suitability prior to use), does not accept pencil.

  • Rated to −29 °C
  • Resistant to ultraviolet light, mildew, chemicals and dirt and won't discolor, crack or tear
  • Excellent for labeling microscopic slides - adhesive backing sticks well to any plastic, glass or metal surface
  • Chemically compatible with most lab reagents although solvents may dissolve adhesive
  • Autoclavable

Formatted for easy setup in word processing programs; instructions included in each pack.

Specifications
  • Colour:
    White
  • Description:
    Labels
  • Dimensions:
    20×38 mm
  • Material:
    Polyolefin
  • Cat. No.:
    817-0082
  • Environmental certification:
    NAR Certificate of Retirement
  • Environmentally Preferable:
    Low Manufacturing Impact
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.