Labels, pressure-sensitive
Labels, Polyolefin, White, 22×22 mm
817-0081
These Nalgene™ polyolefin pressure-sensitive labels are used with photocopiers, laser printers or inkjet printers. Accepts a variety of inks including copier, marker, ballpoint, gels, and technical markers (pretest specific inks for suitability prior to use), does not accept pencil.
- Rated to −29 °C
- Resistant to ultraviolet light, mildew, chemicals and dirt and won't discolor, crack or tear
- Excellent for labeling microscopic slides - adhesive backing sticks well to any plastic, glass or metal surface
- Chemically compatible with most lab reagents although solvents may dissolve adhesive
- Autoclavable
Formatted for easy setup in word processing programs; instructions included in each pack.
- Colour:White
- Description:Labels
- Dimensions:22×22 mm
- Material:Polyolefin
- Cat. No.:817-0081
- Environmental certification:NAR Certificate of Retirement
- Environmentally Preferable:Low Manufacturing Impact
Aanbevelingen
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.



