Order Entry
Belgium
Orders LinkContactUsLinkComponent
Bath fluids

Bath fluids

Thermal H250S, 20...250 °C, 274 °C, 230 °C, Light brown, 5 L
Catalog # 461-0478
Supplier:   JULABO GmbH
Bath fluids
Catalog # 461-0478
Supplier:  JULABO GmbH
CAS Number:  
Bath fluids
Bath fluids

Thermal H250S, 20...250 °C, 274 °C, 230 °C, Light brown, 5 L

CAS Number:  
Price
498,00 €
Log in for availability
1 * 5 L
Product Details & Documents

Bath fluids have been carefully selected and proven in long-term service. They are suitable for use with heating and refrigerated circulators whenever water cannot be employed as bath fluid. The choice of bath fluid is critical for best results in temperature control and operation of the circulator.

  • Low toxicity and almost odourless
  • High stability and good thermal conductivity
  • Low viscosity and low corrosiveness
Specifications

Specifications

  • Pk:
    5 L
  • Colour:
    Light brown
  • Fire point:
    274 °C
  • Flash point:
    230 °C
  • Temperature range:
    20...250 °C
  • Type:
    Thermal H250S

Specifications

Aanbevelingen

Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.

Anders ontvangt u algemene aanbevelingen.