Orders LinkContactUsLinkComponent
Compact automatic burettes, Schellbach, BLAUBRAND®
Compact automatic burettes, Schellbach, BLAUBRAND®
25 ml, 0,05 ml, 785 mm, 0,03 ± ml
Catalog # 612-7171
Supplier:   Brand
Compact automatic burettes, Schellbach, BLAUBRAND®
Catalog # 612-7171
Supplier:  Brand
Compact automatic burettes, Schellbach, BLAUBRAND®
Compact automatic burettes, Schellbach, BLAUBRAND®
25 ml, 0,05 ml, 785 mm, 0,03 ± ml
Price
153,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Borosilicate glass 3.3 bottle, class AS, DE-M marking.
  • Calibrated to deliver 'TD, Ex'
  • Schellbach stripe
  • Detachable stopcock
Certifications: DIN EN ISO 385
Ordering information: Please order burette clamps, supports and rubber bellows separately.
Delivery information: Supplied as burette tube, stopcock with precision tip, filling tube (PVC) and 1000 ml PE bottle
Specifications
  • Capacity:
    25 ml
  • Length:
    785 mm
  • Division:
    0,05 ml
  • Tolerance:
    0,03 ± ml
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Single bulb rubber bellows, orange coloured, with valve and tubing
    Single bulb rubber bellows, orange coloured, with valve and tubing
    BRND23400
    Log in for availability
    1 * 1 ST - 17,50 €
Product Information
  • Catalog #
    Capacity
    Availability
    Price
  • Bases for bottles
    Bases for bottles
    BRND23728
    1000 ml
    Log in for availability
    1 * 1 ST - 8,60 €
Product Information
  • Catalog #
    Availability
    Price
  • Spare filling tube for all compact automatic burettes, PVC
    Spare filling tube for all compact automatic burettes, PVC
    BRND11525
    Log in for availability
    1 * 1 ST - 4,80 €
Product Information
  • Catalog #
    Capacity
    Availability
    Price
  • Bases for bottles
    Bases for bottles
    BRND23725
    500 ml
    Log in for availability
    1 * 1 ST - 7,20 €
Product Information
  • Catalog #
    Length
    Availability
    Price
  • Pump head, clear, NS 29/32, fitting length 185 mm, nozzle outer Ø 7 mm, Boro 3.3
    Pump head, clear, NS 29/32, fitting length 185 mm, nozzle outer Ø 7 mm, Boro 3.3
    BRND23875
    185 mm
    Log in for availability
    1 * 1 ST - 44,70 €
Product Information
  • Catalog #
    Availability
    Price
  • Spare burette tip, clear borosilicate 3.3 glass, for 10 ml compact burettes and compact automatic burettes
    Spare burette tip, clear borosilicate 3.3 glass, for 10 ml compact burettes and compact automatic burettes
    BRND11510
    Log in for availability
    1 * 1 ST - 6,90 €
Product Information
  • Catalog #
    Availability
    Price
  • Spare burette stopcock, PTFE
    Spare burette stopcock, PTFE
    BRND11805
    Log in for availability
    1 * 1 ST - 50,20 €
Product Information
  • Catalog #
    Availability
    Price
  • Spare burette tip, clear borosilicate 3.3 glass, for 50 ml compact burettes and compact automatic burettes
    Spare burette tip, clear borosilicate 3.3 glass, for 50 ml compact burettes and compact automatic burettes
    BRND11512
    Log in for availability
    1 * 1 ST - 6,90 €
Product Information
  • Catalog #
    Capacity
    Division
    Availability
    Price
  • Spare burette tubes, AR-Glas®
    Spare burette tubes, AR-Glas®
    BRND23838
    50 ml
    0,1 ml
    Log in for availability
    1 * 1 ST - 54,90 €
Product Information
  • Catalog #
    Availability
    Price
  • Spare burette tip, clear borosilicate 3.3 glass, for 25 ml compact burettes and compact automatic burettes
    Spare burette tip, clear borosilicate 3.3 glass, for 25 ml compact burettes and compact automatic burettes
    BRND11511
    Log in for availability
    1 * 1 ST - 6,90 €
Product Information
  • Catalog #
    Capacity
    Division
    Availability
    Price
  • Spare burette tubes, AR-Glas®
    Spare burette tubes, AR-Glas®
    BRND23833
    10 ml
    0,05 ml
    Log in for availability
    1 * 1 ST - 54,90 €
Product Information
  • Catalog #
    Capacity
    Division
    Availability
    Price
  • Spare burette tubes, AR-Glas®
    Spare burette tubes, AR-Glas®
    BRND23836
    25 ml
    0,1 ml
    Log in for availability
    1 * 1 ST - 54,90 €
Product Information
  • Catalog #
    Capacity
    Availability
    Price
  • Reservoir bottle, LDPE, GL 25
    Reservoir bottle, LDPE, GL 25
    BRND129055
    500 ml
    Log in for availability
    1 * 1 ST - 4,50 €
Product Information
  • Catalog #
    Capacity
    Availability
    Price
  • Reservoir bottle, LDPE, GL 28
    Reservoir bottle, LDPE, GL 28
    BRND129060
    1000 ml
    Log in for availability
    1 * 1 ST - 8,00 €