Orders LinkContactUsLinkComponent
Pipette tips, Matrix™ SerialMate™
Pipette tips, Matrix™ SerialMate™
Standard, 0,5 - 30 µl, +, -, 48,26 mm, Matrix™ SerialMate™, 20 racks, 96 each
Catalog # MATR5572
   Low Manufacturing Impact
Pipette tips, Matrix™ SerialMate™
Catalog # MATR5572
   Low Manufacturing Impact
Pipette tips, Matrix™ SerialMate™
Standard, 0,5 - 30 µl, +, -, 48,26 mm, Matrix™ SerialMate™, 20 racks, 96 each
Price
299,00 €
Log in for availability
1 * 1.920 ST
Product Details & Documents
Pipette tips for use with the Matrix™ SerialMate™ pipetting station. Compatible with 96-well plates.
Specifications
  • Packed:
    20 racks, 96 each
  • Sterile:
    +
  • Volume:
    0,5 - 30 µl
  • Length:
    48,26 mm
  • Tip style:
    Standard
  • For use with:
    Matrix™ SerialMate™
  • Filter:
    -
  • Cat. No.:
    MATR5572
  • Environmental certification:
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • Environmentally Preferable:
    Low Manufacturing Impact