Order Entry
Belgium
Orders LinkContactUsLinkComponent
Inserts for storage tubes, 2D Barcoded, Matrix®

Inserts for storage tubes, 2D Barcoded, Matrix®

Insert for 0,5 ml and 1,0 ml screw top tube caps, White
Catalog # 479-1343
   Low Manufacturing Impact
Inserts for storage tubes, 2D Barcoded, Matrix®
Catalog # 479-1343
   Low Manufacturing Impact
Inserts for storage tubes, 2D Barcoded, Matrix®

Insert for 0,5 ml and 1,0 ml screw top tube caps, White

Price
38,60 €
Log in for availability
1 * 500 ST
Product Details & Documents

Color inserts snap into cap for a secure fit.

  • Inserts are removable
  • Free of DNA, DNAse, RNAse, PCR Inhibitors, endotoxins and cytotoxins
Specifications
  • Colour:
    White
  • Description:
    Insert for 0,5 ml and 1,0 ml screw top tube caps
  • Seq:
    02
  • Cat. No.:
    479-1343
  • Environmental certification:
    NAR Certificate of Retirement
  • Environmentally Preferable:
    Low Manufacturing Impact
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.