ContactUsLinkComponent
1.4mL V-bottom tubes, sterile, 2D-barcode, TrakMates. Racked in polycarbonate snap racks. V-bottom design enhances small volume flui d recovery. 10 racks of 96 tubes per case
Catalog # 479-1064
1.4mL V-bottom tubes, sterile, 2D-barcode, TrakMates. Racked in polycarbonate snap racks. V-bottom design enhances small volume flui d recovery. 10 racks of 96 tubes per case
Catalog # 479-1064