Orders LinkContactUsLinkComponent
Overflow volumetric flasks, class B
Overflow volumetric flasks, class B
43,5 ml
Catalog # HECH42115012
Overflow volumetric flasks, class B
Catalog # HECH42115012
Overflow volumetric flasks, class B
Overflow volumetric flasks, class B
43,5 ml
Price
22,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
DURAN® borosilicate glass 3.3.
  • Acid-proof and alkali-resistant markings
  • Very good temperature resistance
  • Very good chemical resistance
For water analysis.
Specifications
  • Capacity:
    43,5 ml
  • Seq:
    02