Orders LinkContactUsLinkComponent
Overflow volumetric flasks, class B
Overflow volumetric flasks, class B
250 ml
  HECH42115048
Overflow volumetric flasks, class B
  HECH42115048
Overflow volumetric flasks, class B
Overflow volumetric flasks, class B
250 ml
28,00 €
1 * 1 ST
DURAN® borosilicate glass 3.3.
  • Acid-proof and alkali-resistant markings
  • Very good temperature resistance
  • Very good chemical resistance
For water analysis.
  • Capacity:
    250 ml
  • Seq:
    07