Orders LinkContactUsLinkComponent
Overflow volumetric flasks, class B
Overflow volumetric flasks, class B
164 ml
Catalog # HECH42115042
Overflow volumetric flasks, class B
Catalog # HECH42115042
Overflow volumetric flasks, class B
Overflow volumetric flasks, class B
164 ml
Price
33,50 €
Log in for availability
1 * 1 ST
Product Details & Documents
DURAN® borosilicate glass 3.3.
  • Acid-proof and alkali-resistant markings
  • Very good temperature resistance
  • Very good chemical resistance
For water analysis.
Specifications
  • Capacity:
    164 ml
  • Seq:
    06