Order Entry
Belgium
Orders LinkContactUsLinkComponent
Tank liners, HyPerforma™

Tank liners, HyPerforma™

Tank liners, with liner Ø 630 mm, 200 l
Catalog # HYCPSH30647.03
Tank liners, HyPerforma™
Catalog # HYCPSH30647.03
Tank liners, HyPerforma™

Tank liners, with liner Ø 630 mm, 200 l

Price
1 010,00 €
Log in for availability
1 * 10 ST
Product Details & Documents

Tank liners are gamma irradiated, open single use bags designed to fit in cylindrical drums.

  • Gusset design allows for a close fit to the tank
  • Constructed of CX3-9 film
  • For HyPerforma™ Single-Use Mixer DS 300
Specifications
  • Description:
    Tank liners, with liner Ø 630 mm, 200 l
  • Seq:
    08