Order Entry
Belgium
Orders LinkContactUsLinkComponent
Buffer solutions, colour coded, Fluka™

Buffer solutions, colour coded, Fluka™

10,00, Violet, 20 °C, Plastic bottle, 6×0,5 L, 3.000 mL
Catalog # 33668-6X500ML
Supplier:   Honeywell Chemicals
Buffer solutions, colour coded, Fluka™
Catalog # 33668-6X500ML
CAS Number:  
Buffer solutions, colour coded, Fluka™

10,00, Violet, 20 °C, Plastic bottle, 6×0,5 L, 3.000 mL

CAS Number:  
Price
205,00 €
Log in for availability
1 * 3.000 mL
Product Details & Documents

These buffer solutions are colour-coded for easy identification and the pH 4 and 7 version have fungicide additive.

Documents

Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    3.000 mL
  • Reference temperature:
    20 °C
  • Colour:
    Violet
  • Pk Info:
    6×0,5 L
  • pH value:
    10,00

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.