Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Buffer solutions, colour coded, Fluka™

10,00, Violet, 20 °C, Plastic bottle, 6×1 L, 6 L
Catalog # 33668-6X1L
Supplier:   Honeywell Chemicals
Buffer solutions, colour coded, Fluka™
Catalog # 33668-6X1L
CAS Number:  
Buffer solutions, colour coded, Fluka™

10,00, Violet, 20 °C, Plastic bottle, 6×1 L, 6 L

CAS Number:  
Price
420,00 €
Log in for availability
1 * 6 L
Product Details & Documents

These buffer solutions are colour-coded for easy identification and the pH 4 and 7 version have fungicide additive.

Documents

Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    6 L
  • Reference temperature:
    20 °C
  • Colour:
    Violet
  • Pk Info:
    6×1 L
  • pH value:
    10,00

Specifications