Order Entry
Belgium
Orders LinkContactUsLinkComponent
Buffer solutions, colour coded, Fluka™

Buffer solutions, colour coded, Fluka™

9,00, Blue, 20 °C, Plastic bottle, 6×1 L, 6 L
Catalog # 33667-6X1L
Supplier:   Honeywell Chemicals
Buffer solutions, colour coded, Fluka™
Catalog # 33667-6X1L
CAS Number:  
Buffer solutions, colour coded, Fluka™

9,00, Blue, 20 °C, Plastic bottle, 6×1 L, 6 L

CAS Number:  
Price
451,00 €
Log in for availability
1 * 6 L
Product Details & Documents

These buffer solutions are colour-coded for easy identification and the pH 4 and 7 version have fungicide additive.

Documents

Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    6 L
  • Reference temperature:
    20 °C
  • Colour:
    Blue
  • Pk Info:
    6×1 L
  • pH value:
    9,00

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.