Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Buffer solutions, colour coded, Fluka™

7,00, Green, 20 °C, Plastic bottle, 6×1 L, 6 L
  33666-6X1L
Buffer solutions, colour coded, Fluka™
  33666-6X1L
 :  
Buffer solutions, colour coded, Fluka™

7,00, Green, 20 °C, Plastic bottle, 6×1 L, 6 L

 :  
 
456,00 €
   
1 * 6 L
 

These buffer solutions are colour-coded for easy identification and the pH 4 and 7 version have fungicide additive.

 

 

 

  • Pack type:
    Plastic bottle
  • Pk:
    6 L
  • Reference temperature:
    20 °C
  • Colour:
    Green
  • Pk Info:
    6×1 L
  • pH value:
    7,00

 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.