Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Buffer solutions, colour coded, Fluka™

Buffer solutions, colour coded, Fluka™

7,00, vert, 20 °C, VOLPAK container, 5 L, 5 L
Catalog # 33666-5L-VP
Supplier:   Honeywell Chemicals
Buffer solutions, colour coded, Fluka™
Catalog # 33666-5L-VP
CAS Number:  
Buffer solutions, colour coded, Fluka™

7,00, vert, 20 °C, VOLPAK container, 5 L, 5 L

CAS Number:  
Price
131,00 €
Log in for availability
1 * 5 L
Product Details & Documents

These buffer solutions are colour-coded for easy identification and the pH 4 and 7 version have fungicide additive.

Documents

Specifications

Specifications

  • Type de conditionnement:
    VOLPAK container
  • Cdt:
    5 L
  • Température de réference:
    20 °C
  • Couleur:
    vert
  • Info cdt:
    5 L
  • Valeur pH:
    7,00

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.