Order Entry
Belgium
Orders LinkContactUsLinkComponent
Buffer solutions, colour coded, Fluka™

Buffer solutions, colour coded, Fluka™

4,00, Red, 20 °C, VOLPAK container, 5 L, 5 L
Catalog # 33665-5L-VP
Supplier:   Honeywell Chemicals
Buffer solutions, colour coded, Fluka™
Catalog # 33665-5L-VP
CAS Number:  
Buffer solutions, colour coded, Fluka™

4,00, Red, 20 °C, VOLPAK container, 5 L, 5 L

CAS Number:  
Price
130,00 €
Log in for availability
1 * 5 L
Product Details & Documents

These buffer solutions are colour-coded for easy identification and the pH 4 and 7 version have fungicide additive.

Documents

Specifications

Specifications

  • Pack type:
    VOLPAK container
  • Pk:
    5 L
  • Reference temperature:
    20 °C
  • Colour:
    Red
  • Pk Info:
    5 L
  • pH value:
    4,00

Specifications