ContactUsLinkComponent
Universal Plan Super Apochromat objective with 10x magnification. With working distance of 3.1 mm, numerical aperture of 0.4 and cover correction of 0.17 mm. 1 * 1 items
Catalog # OLYMN2249100
Supplier:  OLYMPUS OPTICAL
Universal Plan Super Apochromat objective with 10x magnification. With working distance of 3.1 mm, numerical aperture of 0.4 and cover correction of 0.17 mm. 1 * 1 items
Catalog # OLYMN2249100
Supplier:  OLYMPUS OPTICAL
Frequently Bought Together