Order Entry
Belgium
Orders LinkContactUsLinkComponent
Dip slides

Dip slides

Flexible agar-agar contact slides for total flora/yeasts and moulds plus neutraliser, 20
Catalog # LIOF525272
Supplier:   LIOFILCHEM
Dip slides
Catalog # LIOF525272
Supplier:  LIOFILCHEM
CAS Number:  
Dip slides
Dip slides

Flexible agar-agar contact slides for total flora/yeasts and moulds plus neutraliser, 20

CAS Number:  
Price
37,80 €
Log in for availability
1 * 20 ST
Product Details & Documents

Two-sided slide with agar-agar centre. Operating instructions: 1. Unscrew the cap containing the agar-agar slide. Hold the bottle in the other hand and lightly apply pressure to the sampling area using the agar-agar slide; or 2. Apply directly to the middle of the swab brush if you have completed a swab sample; or 3. Immerse the agar-agar slide in the liquid to analyse. 4. Insert the slide into the bottle and screw back on. Place in the oven at 37 °C for 24/48 hours. To detect yeasts, increase the incubation by 24/48 hours at 25/30 °C.

  • Box of 10 slides with separate labels.
  • Use-by date: six months after manufacture
  • Storage: ambient temperature (max. 25 °C)
Specifications

Specifications

  • Pk:
    20
  • Description:
    Flexible agar-agar contact slides for total flora/yeasts and moulds plus neutraliser

Specifications