Orders LinkContactUsLinkComponent
Dip slides
Dip slides
Total / Enterobacteria with neutraliser, 20
Catalog # LIOF525262
Supplier:   LIOFILCHEM
Dip slides
Catalog # LIOF525262
Supplier:  LIOFILCHEM
CAS Number:  
Dip slides
Dip slides
Total / Enterobacteria with neutraliser, 20
CAS Number:  
Price
51,90 €
Connexion Pour la disponibilité
1 * 20 ST
Product Details & Documents
Flexible contact slide for microbiology screening of surfaces.
  • ISO 18593
Documents
Spécifications
Spécifications
  • Pk:
    20
  • Description:
    Total / Enterobacteria with neutraliser
Spécifications