Order Entry
Belgium
Orders LinkContactUsLinkComponent
Dip slides

Dip slides

Flexible agar-agar contact slides for pseudomonas, 20
Catalog # LIOF525282
Supplier:   LIOFILCHEM
Dip slides
Catalog # LIOF525282
Supplier:  LIOFILCHEM
CAS Number:  
Dip slides
Dip slides

Flexible agar-agar contact slides for pseudomonas, 20

CAS Number:  
Price
51,90 €
Log in for availability
1 * 20 ST
Product Details & Documents

Two-sided slide with agar-agar centre. Operating instructions: 1. Unscrew the cap containing the agar-agar slide. Hold the bottle in the other hand and lightly apply pressure to the sampling area using the agar-agar slide; or 2. Apply directly to the middle of the swab brush if you have completed a swab sample; or 3. Immerse the agar-agar slide in the liquid to analyse. 4. Insert the slide into the bottle and screw back on. Place in the oven at 37 °C for 24/48 hours. To detect yeasts, increase the incubation by 24/48 hours at 25/30 °C.

  • Box of 10 slides with separate labels.
  • Use-by date: six months after manufacture
  • Storage: ambient temperature (max. 25 °C)
Specifications

Specifications

  • Pk:
    20
  • Description:
    Flexible agar-agar contact slides for pseudomonas

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.