Orders LinkContactUsLinkComponent
Fork wrenches, BOLA
Fork wrench for thread GL 14/GL 18/GL 25, Glasfibre/polyamide
Catalog # BOHLD647-08
Supplier:   Bohlender
Fork wrenches, BOLA
Catalog # BOHLD647-08
Supplier:  Bohlender
Fork wrenches, BOLA
Fork wrenches, BOLA
Fork wrench for thread GL 14/GL 18/GL 25, Glasfibre/polyamide
Price
121,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
These glass fibre reinforced PA fork wrenches can be used for tightening or opening screw joints also at high working temperatures.
  • Good chemical resistance
  • Temperature resistant from −10 to +140 °C
  • Low weight reduces risk of injury or damage
Specifications
  • Description:
    Fork wrench for thread GL 14/GL 18/GL 25
  • Material:
    Glasfibre/polyamide
  • Seq:
    01