Order Entry
Belgium
Orders LinkContactUsLinkComponent
Storage Boxes with Lid

Storage Boxes with Lid

6 l, Transparent, 340×230×95 mm, SAN
Catalog # SEMA0882
Supplier:   SEMADENI
Storage Boxes with Lid
Catalog # SEMA0882
Supplier:  SEMADENI
Storage Boxes with Lid
Storage Boxes with Lid

6 l, Transparent, 340×230×95 mm, SAN

Price
86,40 €
Log in for availability
1 * 1 ST
Product Details & Documents

These storage boxes with transparent lids are either made of polystyrene (PS) or styrene-acrylonitrile resin (SAN).

Specifications
  • Capacity:
    6 l
  • Colour:
    Transparent
  • Material:
    SAN
  • Seq:
    03
  • W×D×H:
    340×230×95 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.