Orders LinkContactUsLinkComponent
Adapters, Combifix®
Adapters, Combifix®
Luer-Lock (male/male) adapters, Transparent
Catalog # BRAU5206642
Supplier:   B. Braun
Adapters, Combifix®
Catalog # BRAU5206642
Supplier:  B. Braun
Adapters, Combifix®
Adapters, Combifix®
Luer-Lock (male/male) adapters, Transparent
Price
312,00 €
Log in for availability
1 * 100 ST
Product Details & Documents
These adapters are used for connecting fittings.
Suitable for interconnecting male and female fittings with 6% (Luer) and 10% (Rekord) taper fittings.
Documents
Specifications
  • Colour:
    Transparent
  • Description:
    Luer-Lock (male/male) adapters
  • Seq:
    03