Orders LinkContactUsLinkComponent
Forceps, precision
Forceps, precision
Precision forceps, straight, smooth tips, extra sharp, Straight, 130 mm
Catalog # BOCH1950
Supplier:   BOCHEM
Forceps, precision
Catalog # BOCH1950
Supplier:  BOCHEM
Forceps, precision
Forceps, precision
Precision forceps, straight, smooth tips, extra sharp, Straight, 130 mm
Price
64,50 €
Log in for availability
1 * 1 ST
Product Details & Documents
These non serrated forceps are made of durable titanium.
  • High corrosion and thermal resistance
  • Lightweight, high strength
Titanium is resistant to diluted sulphuric acid, hydrochloric acid, solutions containing chloride, cold nitric acid as well as most organic acids and alkaline solutions, such as sodium hydroxide.
Specifications
  • Type:
    Straight
  • Description:
    Precision forceps, straight, smooth tips, extra sharp
  • Length:
    130 mm
  • Seq:
    03