Support clips
Support clip with fitting, O. Ø 85 - 90 mm
Catalog # BOCH596
Supplier: BOCHEM
Jump to:
Product Details & Documents
These support clips are coated with ceramic inside.
- For tubes and cylindrical vessels
- With fitting
Ordering information: The shafts (rods) need to be ordered separately.
Specifications
- Description:Support clip with fitting, O. Ø 85 - 90 mm