ContactUsLinkComponent
Screw joint, for tubing with outer diameter 26mm, thread GL 45, PTFE, screw cap made of PPS, inner parts consisting of a v-ring (PPS ), a tapered ring and a sealing ring (both PTFE) as well as an o-ring (for tubing diameters under 3 mm, not exposed t 1 * 1 items
Catalog # BOHLD632-76
Supplier:  Bohlender
undefined
Screw joint, for tubing with outer diameter 26mm, thread GL 45, PTFE, screw cap made of PPS, inner parts consisting of a v-ring (PPS ), a tapered ring and a sealing ring (both PTFE) as well as an o-ring (for tubing diameters under 3 mm, not exposed t 1 * 1 items
Catalog # BOHLD632-76
Supplier:  Bohlender
Frequently Bought Together