Multi-parameter meter (pH/conductivity), bench, 3540
Bench pH/conductivity meter 3540, EU-plug
Catalog # JENW79000-58
Supplier: Cole-Parmer
Jump to:
Product Details & Documents
Ideal for use in all laboratories where pH and conductivity analyses are required.
- Simultaneous display of pH and conductivity
- Two independent channels to prevent interference between probes
- Additional modes for resistivity, salinity, TDS and mV
- Data logger with auto save/print options
- Storage of up to 500 readings (250 for each mode)
Ordering information: Supplied with glass combination pH electrode 662-0343, glass conductivity probe (K=1, 663-0005), electrode stand and holder, ATC probe 662-0338, BNC shorting plug, pH buffers and power supply.
Specifications
- Description:Bench pH/conductivity meter 3540, EU-plug
- Connections:BNC (pH) and 7–pin DIN (conductivity)
- Compliance:GLP
- Conductivity cell constant:0,010 - 19,999 cm⁻¹
- Conductivity resolution:0,01 µS to 1 mS
- Data logging capacity:Up to 500 readings (250 for each mode)
- mV resolution:0,1
- Conductivity accuracy:±0,5% ±2 digits
- Conductivity calibration:Automatic, 1, 2 or 3 points
Automatic standard recognition: 10 µS, 84 µS, 1413 µS, 12,88 mS (with manual override) - W×D×H:210×250×55 mm
- Temperature range:–10...+105 °C
- mV range:±1999,9
- pH range:–2,000...+20,000
- Temperature compensation:ATC and manual, 0...+100 °C
- pH calibration:Automatic, 1, 2 or 3-point
Auto. buffer recognition: Jenway, NIST, DIN and JIS - Temperature resolution:0,1 °C
- Temperature accuracy:±0,5 °C
- Weight:0,850 kg
- Cat. No.:JENW79000-58
- pH accuracy:±0,003
- pH resolution:0,001 / 0,01 / 0,1 selectable
- Model:3540
- Conductivity range:Auto–selected from 0,01 µS to 1999 mS/cm
- Outputs:Analogue, RS232 and IrDA interface
- mV accuracy:±0,2
- Display:Backlit 1/8 VGA LCD



