Cryogenic tubes
2,00 ml, Round bottom, 12,3×46 mm, Yes
RATI5110120
: RATIOLAB
PP tubes with integrated seal in lid and white writing area. Developed for storing and conserving the tissue culture and biological samples at extremely low temperatures. Suitable for use in freezers as well as in gaseous phase of liquid nitrogen to -190 °C.
- White graduated
- Equipped with base for using with one hand
- Free of DNA, DNase, pyrogens and ATP
Produced in a class 7 cleanroom (class 10000) according to DIN ISO 13485:2004, ISO 14644 and 14698 (Federal Standard 209).
: Produced in a class 7 cleanroom (class 10000) according to DIN ISO 13485:2004, ISO 14644 and 14698 (Federal Standard 209).
: Packaged in PE bags containing 100 tubes in boxes.
- Cap included:Yes
- Ø×H:12,3×46 mm
- Capacity:2,00 ml
- Seq:02
- Base:Round bottom



