Orders LinkContactUsLinkComponent
Cryoboxes
Cryoboxes
-, -, White, 75×75×50 mm
Catalog # RATI5160035
Supplier:   RATIOLAB
Cryoboxes
Catalog # RATI5160035
Supplier:  RATIOLAB
Cryoboxes
Cryoboxes
-, -, White, 75×75×50 mm
Price
44,70 €
Log in for availability
1 * 10 ST
Product Details & Documents
Cryoboxes, cardboard, for storage of cryovials at low temperatures.
  • Waterproof coating
  • Dividers must to be ordered separately
  • Available in different heights and five colours
Specifications
  • Compartments:
    -
  • For:
    -
  • W×D×H ext.:
    75×75×50 mm
  • Colour:
    White