Microplates, 96-well
96 Well plate, Clear, 96, -
Catalog # RATI6018111
Supplier: RATIOLAB
Jump to:
Product Details & Documents
Documents
Specifications
- No. of wells:96
- Sterile:-
- Colour:Clear
- Description:96 Well plate
- Cat. No.:RATI6018111
- Model:Micro Test Plates, volume ∼300 μl
- Well profile:U-bottom