Storage Insert Boxes
Grey, 108×216×45 mm, PS
HUEN622500
Insert boxes made of tough polystyrene (PS) for order in drawers, tool cupboards, storage cabinets and small parts boxes.
- Colours suitable for foods
- Suitable for assortment boxes and small parts cases
: Transparent lids are available separately.
- Colour:Grey
- Material:PS
- Seq:12
- W×D×H:108×216×45 mm
Accessories
- DescriptionColour
- DescriptionColour
- DescriptionColour
- DescriptionColour
- DescriptionColour
- DescriptionColour
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



