ContactUsLinkComponent
KIT CERAMIC 1,4/2,8MM IN 2 ML TUBES 50 PREP. ready-to-use kits with the Precellys24, includes 50 microtubes, prefilled with 1,4 a nd 2,8mm ceramic (zirconium oxide) beads in 2mL bead beating custom tubes, for tissue homogenizing like brain, liver, l 1 * 1 KIT
Catalog # 432-3762
Supplier:  Bertin Instruments
KIT CERAMIC 1,4/2,8MM IN 2 ML TUBES 50 PREP. ready-to-use kits with the Precellys24, includes 50 microtubes, prefilled with 1,4 a nd 2,8mm ceramic (zirconium oxide) beads in 2mL bead beating custom tubes, for tissue homogenizing like brain, liver, l 1 * 1 KIT
Catalog # 432-3762
Supplier:  Bertin Instruments
Frequently Bought Together