Order Entry
Belgium
Orders LinkContactUsLinkComponent
Ammonia buffer solution pH10 (20 °C) for complexometry, Fluka™

Ammonia buffer solution pH10 (20 °C) for complexometry, Fluka™

Ammonia buffer, 10, Plastic bottle, 1 L
Catalog # 33582-1L
Supplier:   Honeywell Chemicals
Ammonia buffer solution pH10 (20 °C) for complexometry, Fluka™
Catalog # 33582-1L
CAS Number:  
Ammonia buffer solution pH10 (20 °C) for complexometry, Fluka™

Ammonia buffer, 10, Plastic bottle, 1 L

CAS Number:  
Price
54,10 €
Log in for availability
1 * 1 L
Product Details & Documents

Documents

Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Description:
    Ammonia buffer
  • pH value:
    10

Specifications