Order Entry
Belgium
Orders LinkContactUsLinkComponent
Tris-Glycine buffer

Tris-Glycine buffer

Tris-glycine buffer (10X for 5 l), Glass bottle, 500 mL
Catalog # EDVO656
Supplier:   EDVOTEK
Tris-Glycine buffer
Catalog # EDVO656
Supplier:  EDVOTEK
CAS Number:  
Tris-Glycine buffer

Tris-glycine buffer (10X for 5 l), Glass bottle, 500 mL

CAS Number:  
Price
27,50 €
Log in for availability
1 * 500 mL
Product Details & Documents

For vertical polyacrylamide gel electrophoresis.

Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Description:
    Tris-glycine buffer (10X for 5 l)

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.