Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Tris-Glycine buffer

Tris-glycine-SDS buffer (10X for 5 l), Glass bottle, 500 mL
Catalog # EDVO655
Supplier:   EDVOTEK
Tris-Glycine buffer
Catalog # EDVO655
Supplier:  EDVOTEK
CAS Number:  
Tris-Glycine buffer

Tris-glycine-SDS buffer (10X for 5 l), Glass bottle, 500 mL

CAS Number:  
Price
27,50 €
Log in for availability
1 * 500 mL
Product Details & Documents

For vertical polyacrylamide gel electrophoresis.

Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Description:
    Tris-glycine-SDS buffer (10X for 5 l)

Specifications