Orders LinkContactUsLinkComponent
Single channel pipettes, variable volume
Single channel pipettes, variable volume
5 - 50 µl
Catalog # 613-1359
Supplier:   EDVOTEK
Single channel pipettes, variable volume
Catalog # 613-1359
Supplier:  EDVOTEK
Single channel pipettes, variable volume
Single channel pipettes, variable volume
5 - 50 µl
Price
105,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Sturdily designed handheld pipettes with volumes ranging from 0,1 to 5000 µl. They are easy to use, accurate and use standard micropipette tips.
  • Volume easily selected by twisting the top
  • Lightweight design and tip ejector makes operation fast and easy
  • Tool and instructions included for self-calibration
Specifications
  • Volume:
    5 - 50 µl