Orders LinkContactUsLinkComponent
Eppendorf® Assay/Reader Microplates
Eppendorf® Assay/Reader Microplates
V-bottom, PCR clean, White/grey, 96, -
N° de catalogue 737-0170
   Low Manufacturing Impact
Fournisseur:   EPPENDORF
Eppendorf® Assay/Reader Microplates
N° de catalogue 737-0170
Fournisseur:  EPPENDORF
   Low Manufacturing Impact
Eppendorf® Assay/Reader Microplates
Eppendorf® Assay/Reader Microplates
V-bottom, PCR clean, White/grey, 96, -
Prix
354,00 €
Connexion Pour la disponibilité
1 * 80 ST
Product Details & Documents
Eppendorf Assay/Reader plates are designed for diverse microplate assays, optimising absorbance, fluorescence, and chemiluminescence. With options for both top- and bottom-reading plate readers, they ensure clear signals even with low-concentration samples.
  • Black Eppendorf Microplates offer an excellent signal-to-noise ratio – for clear signals even with low-concentration samples
  • White Eppendorf Microplates are optimized for highest sensitivity in the detection of luminescence signals by maximising reflection
  • Solid black and white assay plates are made of polypropylene resulting in high resistance to chemicals, mechanical stress, and temperature extremes
  • All plates are optimised for minimal autofluorescence and autoluminescence
  • Available in lot-certified PCR clean purity grade
  • Outstanding dimensional accuracy and high well-to well homogeneity for perfect automation suitability
Perfect plate for your microplate assay protocol. Eppendorf assay plate range has the plate for your application, optimised for measuring absorbance in the visible range, fluorescence and chemiluminescence. The range includes both solid plates for top-reading plate readers and versatile clear and black/clear plates for use with both top- and bottom-reading plate readers.
Experience clear signals even with low-concentration samples. Black microplates are ideal for fluorescence detection, with an excellent signal-to-noise ratio, providing low detection limits in assays. White microplates maximise reflection for high-sensitivity detection of luminescence or weak fluorescence.
Spécifications
  • No. of wells:
    96
  • Sterile:
    -
  • Colour:
    White/grey
  • Description:
    V-bottom, PCR clean
  • Seq:
    2
  • Cat. No.:
    737-0170
  • Environmental certification:
    Renewable Energy Certificate, ISO 14001
  • Environmentally Preferable:
    Low Manufacturing Impact