Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

dNTP sets, illustra™

dNTP set (100 mM each A,C,G,T), 4×25 µmol, 1 SET
Catalog # 28-4065-51
Supplier:   Cytiva
dNTP sets, illustra™
Catalog # 28-4065-51
Supplier:  Cytiva
CAS Number:  
dNTP sets, illustra™
dNTP sets, illustra™

dNTP set (100 mM each A,C,G,T), 4×25 µmol, 1 SET

CAS Number:  
Price
277,00 €
Log in for availability
1 * 1 SET
Product Details & Documents

High purity dNTPs for amplification, dideoxy sequencing, labelling, mutagenesis, cDNA synthesis, and expression profiling.

  • Free from DNase, RNase, and nicking enzyme activity
  • Greater than 99% triphosphate purity (by HPLC) for consistent, high performance
  • Buffer-free, ready to use solutions at a variety of concentrations
  • Functionally tested to produce a 20,7 kb PCR amplification product from lambda DNA
Specifications

Specifications

  • Pk:
    1 SET
  • Description:
    dNTP set (100 mM each A,C,G,T), 4×25 µmol

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.