Order Entry
Belgium
Orders LinkContactUsLinkComponent
PIG® small mobile container kit

PIG® small mobile container kit

Spill kit, includes 100 mat pads, 8 socks, 12 pillows and 5 disposal bags, 80,7 L, 550×480×970 mm
Catalog # 121-0067
Supplier:   NEW PIG
PIG® small mobile container kit
Catalog # 121-0067
Supplier:  NEW PIG
PIG® small mobile container kit
PIG® small mobile container kit

Spill kit, includes 100 mat pads, 8 socks, 12 pillows and 5 disposal bags, 80,7 L, 550×480×970 mm

Price
325,00 €
Log in for availability
1 * 1 KIT
Product Details & Documents

Fast, effective response to indoor and outdoor spills.

  • Durable, wheeled 120 L container is packed with high-quality PIG® absorbents
  • Easy to transport supplies directly to the spill site
  • Hinged lid makes it easy to get the absorbents you need — just flip it open
  • Container with bold Spill Kit lettering for quick identification
  • Absorbents are organised for quick, easy response
  • Heavy-duty rubber wheels roll easily over nearly any surface

The weather-resistant polyethylene container protects the contents from moisture, dirt and damage (containers are best kept indoors for long-term storage).

Caution: To ensure effectiveness and your safety, we recommend that you conduct compatibility and absorption testing of your chemicals with PIG® HAZ-MAT products prior to purchase.

Specifications
  • Description:
    Spill kit, includes 100 mat pads, 8 socks, 12 pillows and 5 disposal bags
  • Seq:
    03
  • W×D×H:
    550×480×970 mm
  • Absorbency:
    80,7 L
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.